Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ERD2
- Entrez Gene ID
-
842094
- Gene Name
- heat shock protein 70 (Hsp 70) family protein
- Gene Aliases
-
AT1G56410, EARLY-RESPONSIVE TO DEHYDRATION 2, F13N6.9, F13N6_9, HEAT SHOCK PROTEIN 70T-1, HSP70T-1
- Chromosome Location
-
Chr.1: 21117147 - 21119241 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104519.1 |
NP_176036.1 |
1 - 2 |
213 |
116 |
GenBank mRNA |
DQ446367.1 |
- |
1 - 2 |
213 |
116 |
View Full Product Details