Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl14
Entrez Gene ID
820403
Gene Name
serine carboxypeptidase-like 14
Gene Aliases
AT3G12230, serine carboxypeptidase-like 14
Chromosome Location
Chr.3: 3899364 - 3901879 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112059.2 NP_187831.1 12 - 13 1178 139

View Full Product Details