Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71B36
- Entrez Gene ID
-
822236
- Gene Name
- cytochrome P450, family 71, subfamily B, polypeptide 36
- Gene Aliases
-
''cytochrome P450, AT3G26320, cytochrome P450, family 71, polypeptide 36, polypeptide 36'', subfamily B
- Chromosome Location
-
Chr.3: 9644319 - 9646093 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113539.2 |
NP_189263.1 |
1 - 2 |
335 |
158 |
View Full Product Details