Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCB13
- Entrez Gene ID
-
839687
- Gene Name
- P-glycoprotein 13
- Gene Aliases
-
AT1G27940, ATP-binding cassette B13, F13K9.5, F13K9_5, P-glycoprotein 13, PGP13
- Chromosome Location
-
Chr.1: 9733430 - 9738236 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102559.2 |
NP_174115.1 |
3 - 4 |
613 |
108 |
View Full Product Details