Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eIF4E3
- Entrez Gene ID
-
839836
- Gene Name
- translation initiation factor
- Gene Aliases
-
AT1G29590, EIF4E1C, F15D2.16, F15D2_16, eukaryotic translation Initiation Factor 4E3
- Chromosome Location
-
Chr.1: 10339992 - 10341832 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102699.2 |
NP_174252.2 |
2 - 3 |
553 |
156 |
View Full Product Details