Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY14
Entrez Gene ID
839945
Gene Name
WRKY DNA-binding protein 14
Gene Aliases
AR411, AT1G30650, ATWRKY14, T5I8.10, T5I8_10, WRKY DNA-BINDING PROTEIN 14, WRKY DNA-binding protein 14
Chromosome Location
Chr.1: 10868218 - 10871297 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102802.2 NP_564359.1 2 - 3 1103 154
GenBank mRNA AK228343.1 - 2 - 3 959 154
BT006468.1 - 2 - 3 709 154

View Full Product Details