Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl37
Entrez Gene ID
824364
Gene Name
serine carboxypeptidase-like 37
Gene Aliases
AT3G52010, serine carboxypeptidase-like 37
Chromosome Location
Chr.3: 19296183 - 19298171 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115060.3 NP_190769.1 2 - 3 515 131

View Full Product Details