Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71B31
- Entrez Gene ID
-
824497
- Gene Name
- cytochrome P450, family 71, subfamily B, polypeptide 31
- Gene Aliases
-
''cytochrome P450, AT3G53300, F4P12.1, cytochrome P450, family 71, polypeptide 31, polypeptide 31'', subfamily B
- Chromosome Location
-
Chr.3: 19760475 - 19762357 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115190.1 |
NP_190898.1 |
1 - 2 |
940 |
130 |
View Full Product Details