Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CPK27
- Entrez Gene ID
-
825805
- Gene Name
- calcium-dependent protein kinase 27
- Gene Aliases
-
AT4G04700, T4B21.21, T4B21_21, calcium-dependent protein kinase 27
- Chromosome Location
-
Chr.4: 2385038 - 2388105 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116708.3 |
NP_192379.2 |
7 - 8 |
1476 |
134 |
View Full Product Details