Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71A19
- Entrez Gene ID
-
826959
- Gene Name
- cytochrome P450, family 71, subfamily A, polypeptide 19
- Gene Aliases
-
''cytochrome P450, AT4G13290, T9E8.30, T9E8_30, cytochrome P450, family 71, polypeptide 19, polypeptide 19'', subfamily A
- Chromosome Location
-
Chr.4: 7740283 - 7743135 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117402.4 |
NP_193065.1 |
1 - 2 |
791 |
148 |
GenBank mRNA |
AY133692.1 |
- |
1 - 2 |
393 |
148 |
View Full Product Details