Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSLC5
Entrez Gene ID
829286
Gene Name
Cellulose-synthase-like C5
Gene Aliases
AT4G31590, ATCSLC05, ATCSLC5, CELLULOSE-SYNTHASE LIKE C5, CSLC05, Cellulose-synthase-like C5, F28M20.220, F28M20_220
Chromosome Location
Chr.4: 15309506 - 15312987 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119308.3 NP_194887.1 3 - 4 1967 123
GenBank mRNA BT003334.1 - 3 - 4 1571 123
BT008423.1 - 3 - 4 1316 123
KJ138731.1 - 3 - 4 1316 123

View Full Product Details