Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUC9
Entrez Gene ID
830504
Gene Name
sucrose-proton symporter 9
Gene Aliases
AT5G06170, ATSUC9, MBL20.5, MBL20_5, sucrose-proton symporter 9
Chromosome Location
Chr.5: 1869791 - 1871719 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120699.1 NP_196235.1 1 - 2 1240 122

View Full Product Details