Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PMSR3
- Entrez Gene ID
-
830638
- Gene Name
- peptidemethionine sulfoxide reductase 3
- Gene Aliases
-
ARABIDOPSIS THALIANA METHIONINE SULFOXIDE REDUCTASE 3, AT5G07470, ATMSRA3, T2I1.180, T2I1_180, peptidemethionine sulfoxide reductase 3
- Chromosome Location
-
Chr.5: 2362524 - 2364479 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120829.3 |
NP_196364.1 |
1 - 2 |
548 |
131 |
GenBank mRNA |
AY044316.1 |
- |
1 - 2 |
355 |
131 |
View Full Product Details