Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUA3
Entrez Gene ID
832097
Gene Name
tubulin alpha-3
Gene Aliases
AT5G19770, T29J13.190, T29J13_190, tubulin alpha-3
Chromosome Location
Chr.5: 6682420 - 6684744 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121982.4 NP_197478.1 1 - 2 378 152
GenBank mRNA AF367301.1 - 1 - 2 208 152
AY065164.1 - 1 - 2 206 152
AY143895.1 - 1 - 2 108 152
BT000718.1 - 1 - 2 206 152
BT000719.1 - 1 - 2 206 152
BT001197.1 - 1 - 2 108 152

View Full Product Details