Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GolS5
Entrez Gene ID
832444
Gene Name
galactinol synthase 5
Gene Aliases
AT5G23790, AtGolS5, MRO11.17, MRO11_17, galactinol synthase 5
Chromosome Location
Chr.5: 8019953 - 8021650 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122284.2 NP_197768.1 2 - 3 699 87

View Full Product Details