Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71B11
- Entrez Gene ID
-
832583
- Gene Name
- cytochrome p450, family 71, subfamily B, polypeptide 11
- Gene Aliases
-
''ytochrome p450, AT5G25120, T11H3.130, T11H3_130, family 71, polypeptide 11, polypeptide 11'', subfamily B, ytochrome p450
- Chromosome Location
-
Chr.5: 8661970 - 8664681 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122421.3 |
NP_197894.1 |
2 - 3 |
1154 |
144 |
GenBank mRNA |
AK176623.1 |
- |
2 - 3 |
1025 |
144 |
View Full Product Details