Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ADF3
- Entrez Gene ID
-
836110
- Gene Name
- actin depolymerizing factor 3
- Gene Aliases
-
ACTIN DEPOLYMERIZING FACTOR 3, AT5G59880, MMN10.4, MMN10_4, actin depolymerizing factor 3
- Chromosome Location
-
Chr.5: 24120164 - 24121949 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125381.2 |
NP_568915.2 |
2 - 3 |
420 |
95 |
View Full Product Details