Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUM18
Entrez Gene ID
836133
Gene Name
pumilio 18
Gene Aliases
APUM18, AT5G60110, MGO3.9, MGO3_9, pumilio 18
Chromosome Location
Chr.5: 24201954 - 24202937 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125404.1 NP_200819.1 1 - 1 942 137
GenBank mRNA DQ056729.1 - 1 - 1 942 137

View Full Product Details