Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLK3
Entrez Gene ID
828656
Gene Name
SEUSS-like 3
Gene Aliases
AT4G25515, SEUSS-like 3
Chromosome Location
Chr.4: 13028219 - 13031803 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148375.6 NP_680741.5 6 - 7 1102 98

View Full Product Details