Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chr31
Entrez Gene ID
837051
Gene Name
chromatin remodeling 31
Gene Aliases
AT1G05490, T25N20.14, chromatin remodeling 31
Chromosome Location
Chr.1: 1618504 - 1623489 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100428.4 NP_172040.2 2 - 2 2291 82

View Full Product Details