Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AT1G07660
- Entrez Gene ID
-
837279
- Gene Name
- Histone superfamily protein
- Gene Aliases
-
AT1G07660, F24B9.25
- Chromosome Location
-
Chr.1: 2368833 - 2369735 on Build TAIR10.1
- Gene Symbol
- HIS4
- Entrez Gene ID
-
817423
- Gene Name
- histone H4
- Gene Aliases
-
AT2G28740, F8N16.2, F8N16_2, histone H4
- Chromosome Location
-
Chr.2: 12329345 - 12330059 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK175513.1 |
- |
2 - 2 |
436 |
139 |
AK175843.1 |
- |
2 - 2 |
560 |
139 |
AK176147.1 |
- |
2 - 2 |
560 |
139 |
View Full Product Details