Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MDH
Entrez Gene ID
823906
Gene Name
malate dehydrogenase
Gene Aliases
AT3G47520, MALATE DEHYDROGENASE, malate dehydrogenase, pNAD-MDH, plastidic NAD-dependent malate dehydrogenase
Chromosome Location
Chr.3: 17512816 - 17515155 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114620.3 NP_190336.1 2 - 2 1919 183
GenBank mRNA AY128281.1 - 2 - 2 1516 183

View Full Product Details