Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SZF1
Entrez Gene ID
824764
Gene Name
salt-inducible zinc finger 1
Gene Aliases
AT3G55980, ATSZF1, salt-inducible zinc finger 1
Chromosome Location
Chr.3: 20776220 - 20778952 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339762.1 NP_001326199.1 2 - 2 2171 120
NM_115456.3 NP_567030.1 2 - 2 2126 120
GenBank mRNA AY056282.1 - 2 - 2 2098 120
BT008625.1 - 2 - 2 2035 120

View Full Product Details