Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBQ10
Entrez Gene ID
825880
Gene Name
polyubiquitin 10
Gene Aliases
AT4G05320, C17L7.240, C17L7_240, UBI10, polyubiquitin 10, ubiquitin 10
Chromosome Location
Chr.4: 2717977 - 2720308 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084884.5 NP_001078353.4 3 - 3 1568 120
NM_001340546.1 NP_001328999.1 3 - 3 694 120
NM_116771.5 NP_567291.4 3 - 3 1378 120
NM_178968.5 NP_849299.4 2 - 2 1799 120
NM_178970.5 NP_849301.4 2 - 2 1571 120
NM_202787.4 NP_974516.4 3 - 3 1150 120
GenBank mRNA AY139999.1 - 2 - 2 1587 120

View Full Product Details