Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOB
Entrez Gene ID
836429
Gene Name
Lateral organ boundaries (LOB) domain family protein
Gene Aliases
AT5G63090, LATERAL ORGAN BOUNDARIES, MDC12.5, MDC12_5
Chromosome Location
Chr.5: 25308516 - 25311019 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125703.4 NP_201114.1 2 - 2 683 66
NM_180921.2 NP_851252.1 2 - 2 633 66
NM_180922.1 NP_851253.1 3 - 3 692 66
NM_180923.1 NP_851254.1 3 - 3 763 66
GenBank mRNA AB164305.1 - 1 - 1 319 66
AB473837.1 - 2 - 2 608 66
AF447897.1 - 2 - 2 633 66
BT025745.1 - 1 - 1 319 66

View Full Product Details