Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR69
Entrez Gene ID
814741
Gene Name
low-molecular-weight cysteine-rich 69
Gene Aliases
AT2G02100, F5O4.13, F5O4_13, PDF2.2, low-molecular-weight cysteine-rich 69
Chromosome Location
Chr.2: 528179 - 529186 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126271.3 NP_178319.1 2 - 2 582 149
GenBank mRNA AF370355.1 - 2 - 2 411 149

View Full Product Details