Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHT1;4
Entrez Gene ID
818479
Gene Name
phosphate transporter 1;4
Gene Aliases
ARABIDOPSIS THALIANA PHOSPHATE TRANSPORTER 2, AT2G38940, ATPT2, T7F6.11, T7F6_11, phosphate transporter 1;4
Chromosome Location
Chr.2: 16257157 - 16260253 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129452.4 NP_181428.1 2 - 2 1835 125
GenBank mRNA U62331.1 - 2 - 2 1741 125

View Full Product Details