Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDF2
Entrez Gene ID
821819
Gene Name
prefoldin 2
Gene Aliases
AT3G22480, prefoldin 2
Chromosome Location
Chr.3: 7967989 - 7969705 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113147.4 NP_188887.1 2 - 2 663 146
NM_180295.1 NP_850626.1 2 - 2 472 146
GenBank mRNA AK221809.1 - 2 - 2 574 146
AY034920.1 - 2 - 2 557 146
AY086635.1 - 2 - 2 514 146

View Full Product Details