Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZAR1
Entrez Gene ID
824259
Gene Name
HOPZ-ACTIVATED RESISTANCE 1
Gene Aliases
AT3G50950, HOPZ-ACTIVATED RESISTANCE 1
Chromosome Location
Chr.3: 18935674 - 18938799 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114955.4 NP_190664.1 2 - 2 1369 75
NM_180346.1 NP_850677.1 2 - 2 1335 75
GenBank mRNA AK227017.1 - 2 - 2 1218 75

View Full Product Details