Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TUA2
- Entrez Gene ID
-
841425
- Gene Name
- tubulin alpha-2 chain
- Gene Aliases
-
AT1G50010, F2J10.11, F2J10_11, tubulin alpha-2 chain
- Chromosome Location
-
Chr.1: 18517586 - 18520000 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103889.3 |
NP_175423.1 |
3 - 3 |
788 |
128 |
- Gene Symbol
- TUA4
- Entrez Gene ID
-
839405
- Gene Name
- tubulin alpha-4 chain
- Gene Aliases
-
AT1G04820, F13M7.19, TOR2, TORTIFOLIA 2, tubulin alpha-4 chain
- Chromosome Location
-
Chr.1: 1356084 - 1358547 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF410330.1 |
- |
3 - 3 |
758 |
128 |
AY065117.1 |
- |
3 - 3 |
760 |
128 |
AY149922.1 |
- |
3 - 3 |
637 |
128 |
View Full Product Details