Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCJ18
Entrez Gene ID
3768365
Gene Name
cyclin J18
Gene Aliases
AT2G01905, cyclin J18
Chromosome Location
Chr.2: 410321 - 416000 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035867.3 NP_001030944.1 2 - 3 181 112

View Full Product Details