Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCRL9
Entrez Gene ID
3768317
Gene Name
SCR-like 9
Gene Aliases
AT2G05117, SCR-like 9
Chromosome Location
Chr.2: 1838592 - 1839014 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036247.1 NP_001031324.1 1 - 2 65 81

View Full Product Details