Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR81
Entrez Gene ID
3767828
Gene Name
low-molecular-weight cysteine-rich 81
Gene Aliases
AT2G25295, low-molecular-weight cysteine-rich 81
Chromosome Location
Chr.2: 10769877 - 10770485 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036335.2 NP_001031412.1 1 - 2 122 88

View Full Product Details