Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR37
Entrez Gene ID
3770153
Gene Name
low-molecular-weight cysteine-rich 37
Gene Aliases
AT4G13095, low-molecular-weight cysteine-rich 37
Chromosome Location
Chr.4: 7634930 - 7635673 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036546.2 NP_001031623.1 1 - 2 111 77

View Full Product Details