Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR15
Entrez Gene ID
3770495
Gene Name
low-molecular-weight cysteine-rich 15
Gene Aliases
AT4G19038, low-molecular-weight cysteine-rich 15
Chromosome Location
Chr.4: 10429871 - 10431042 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036592.3 NP_001031669.1 1 - 2 143 82

View Full Product Details