Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR63
Entrez Gene ID
3770559
Gene Name
low-molecular-weight cysteine-rich 63
Gene Aliases
AT4G30067, low-molecular-weight cysteine-rich 63
Chromosome Location
Chr.4: 14696074 - 14696597 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036677.2 NP_001031754.1 1 - 2 65 104

View Full Product Details