Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TIF3B1
- Entrez Gene ID
-
832826
- Gene Name
- translation initiation factor 3B1
- Gene Aliases
-
ARABIDOPSIS THALIANA TRANSLATION INITIATION FACTOR 3B1, AT5G27640, ATEIF3B-1, ATTIF3B1, EIF3B, EIF3B-1, EUKARYOTIC TRANSLATION INITIATION FACTOR 3B, EUKARYOTIC TRANSLATION INITIATION FACTOR 3B1, T1G16.2, translation initiation factor 3B1
- Chromosome Location
-
Chr.5: 9780609 - 9785288 on Build TAIR10.1
View Full Product Details