Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR57
Entrez Gene ID
3771402
Gene Name
low-molecular-weight cysteine-rich 57
Gene Aliases
AT5G42242, low-molecular-weight cysteine-rich 57
Chromosome Location
Chr.5: 16891188 - 16891909 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036919.1 NP_001031996.1 1 - 2 65 88

View Full Product Details