Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOPP8
Entrez Gene ID
832846
Gene Name
Calcineurin-like metallo-phosphoesterase superfamily protein
Gene Aliases
AT5G27840, T1G16.170, T1G16_170
Chromosome Location
Chr.5: 9862923 - 9865172 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344043.1 NP_001330891.1 3 - 4 1422 122
NM_122666.4 NP_568501.3 4 - 5 1237 122

View Full Product Details