Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLP28
Entrez Gene ID
843420
Gene Name
MLP-like protein 28
Gene Aliases
AT1G70830, F15H11.8, F15H11_8, MLP-like protein 28
Chromosome Location
Chr.1: 26709889 - 26711587 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105751.5 NP_177240.3 2 - 3 806 121

View Full Product Details