Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR53
Entrez Gene ID
3769757
Gene Name
low-molecular-weight cysteine-rich 53
Gene Aliases
AT3G61177, low-molecular-weight cysteine-rich 53
Chromosome Location
Chr.3: 22644364 - 22644819 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035834.2 NP_001030911.1 1 - 2 70 132

View Full Product Details