Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIL3:1
Entrez Gene ID
827479
Gene Name
Chlorophyll A-B binding family protein
Gene Aliases
AT4G17600, DL4835W, FCAALL.30
Chromosome Location
Chr.4: 9803624 - 9805410 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117868.6 NP_567532.1 1 - 2 530 145
GenBank mRNA AF134133.1 - 1 - 2 400 145
AF375427.1 - 1 - 2 428 145
AY074821.1 - 1 - 2 395 145
AY136323.1 - 1 - 2 448 145
BT000402.1 - 1 - 2 395 145
BT000425.1 - 1 - 2 448 145
BT002185.1 - 1 - 2 395 145

View Full Product Details