Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GF14 PHI
Entrez Gene ID
840403
Gene Name
GF14 protein phi chain
Gene Aliases
14-3-3 PROTEIN G-BOX FACTOR14 PHI, 14-3-3PHI, AT1G35160, GENERAL REGULATORY FACTOR 4, GF14 protein phi chain, GRF4, T32G9.30, T32G9_30
Chromosome Location
Chr.1: 12867081 - 12868890 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198209.1 NP_001185138.1 2 - 3 754 136
NM_103213.3 NP_564453.1 2 - 3 799 136
GenBank mRNA AY084342.1 - 2 - 3 699 136
L09111.1 - 2 - 3 667 136

View Full Product Details