Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS12
Entrez Gene ID
826958
Gene Name
terpenoid synthase 12
Gene Aliases
AT4G13280, ATTPS12, T9E8.20, T9E8_20, terpenoid synthase 12
Chromosome Location
Chr.4: 7733114 - 7736745 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117401.2 NP_193064.2 6 - 7 1179 92

View Full Product Details