Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YAB2
Entrez Gene ID
3766682
Gene Name
Plant-specific transcription factor YABBY family protein
Gene Aliases
AT1G08465, T27G7.15, YABBY2
Chromosome Location
Chr.1: 2675813 - 2679824 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084021.2 NP_001077490.1 4 - 5 598 97
GenBank mRNA AF136539.1 - 4 - 5 404 97

View Full Product Details