Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIR
Entrez Gene ID
843624
Gene Name
toll/interleukin-1 receptor-like protein
Gene Aliases
AT1G72930, AtTN10, F3N23.13, F3N23_13, TIR-nucleotide binding site family 10, TN10, toll/interleukin-1 receptor-like
Chromosome Location
Chr.1: 27439240 - 27440464 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084347.1 NP_001077816.1 1 - 2 524 136

View Full Product Details