Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSBX
Entrez Gene ID
815210
Gene Name
photosystem II subunit X
Gene Aliases
AT2G06520, PSII-X, T12H3.7, T12H3_7, photosystem II subunit X
Chromosome Location
Chr.2: 2587729 - 2588448 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126649.3 NP_565335.1 1 - 1 446 96
GenBank mRNA AF370573.1 - 1 - 1 216 96
AK226390.1 - 1 - 1 276 96

View Full Product Details