Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTFL7
Entrez Gene ID
28719406
Gene Name
ROTUNDIFOLIA like 7
Gene Aliases
AT3G55515, DEVIL 8, DVL8, ROTUNDIFOLIA like 7
Chromosome Location
Chr.3: 20590263 - 20591401 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339732.1 NP_001319758.1 1 - 1 711 74

View Full Product Details