Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEP7
Entrez Gene ID
5008207
Gene Name
elicitor peptide 7 precursor
Gene Aliases
AT5G09978, elicitor peptide 7 precursor
Chromosome Location
Chr.5: 3120655 - 3121879 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085092.3 NP_001078561.1 1 - 2 205 93
GenBank mRNA DQ487624.1 - 1 - 2 132 93

View Full Product Details