Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC17A6
Entrez Gene ID
100125225
Gene Name
solute carrier family 17 member 6
Gene Aliases
VGluT2
Chromosome Location
Chr.29: 22471039 - 22512128 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103110.1 NP_001096580.1 2 - 3 451 76
GenBank mRNA BC147973.1 - 2 - 3 451 76

View Full Product Details